(S)-4-Cyano-4'-(2-methylbutyl)biphenyl ≥98.0% (by GC)

Supplier: TCI America
Warning

Synonyms: (S)-4'-(2-Methylbutyl)-[1,1'-biphenyl]-4-carbonitrile

C2913-1G C2913-5G
TCC2913-1GEA 161.16 CAD
TCC2913-1G TCC2913-5G
(S)-4-Cyano-4'-(2-methylbutyl)biphenyl ≥98.0% (by GC)
(S)-4-Cyano-4'-(2-methylbutyl)biphenyl
Formula: C₁₈H₁₉N
MDL Number: MFCD00192280
CAS Number: 63799-11-1
EINECS: 264-464-8

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +42.0 to +46.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR