(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl ≥98.0% (by GC)

Supplier: TCI America

B3202-1G B3202-5G
TCB3202-1GEA 218.04 CAD
TCB3202-1G TCB3202-5G
(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl ≥98.0% (by GC)
(R)-(+)-2,2'-Bis(methoxymethoxy)-1,1'-binaphthyl
Formula: C₂₄H₂₂O₄
MDL Number: MFCD03788935
CAS Number: 173831-50-0

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Melting point 100.0 to 104.0 deg-C
Specific rotation [a]20/D +91.0 to +95.0 deg (C=1,CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR