(2-Bromophenylethynyl)trimethylsilane ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
((2-Bromophenyl)ethynyl)trimethylsilane
B5138-1G
B5138-5G
76219-504EA
47.4
CAD
76219-504
76219-506
(2-Bromophenylethynyl)trimethylsilane ≥98.0% (by GC)
(2-Bromophenylethynyl)trimethylsilane
Formula:
C₁₁H₁₃BrSi MW: 253.21 g/mol Boiling Pt: 91°C Storage Temperature: Refrigerator |
MDL Number:
MFCD01321391 CAS Number: 38274-16-7 |
Specification Test Results
Appearance | Light yellow to Brown clear liquid |
Purity (GC) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...