Trimethobenzamide hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

T3525-1G T3525-5G
TCT3525-1GEA 169.06 CAD
TCT3525-1G TCT3525-5G
Trimethobenzamide hydrochloride ≥98.0% (by HPLC, titration analysis)
Trimethobenzamide hydrochloride
Formula: C₂₁H₂₈N₂O₅·HCl
Storage Temperature: Refrigerator
MDL Number: MFCD00057999
CAS Number: 554-92-7
EINECS: 209-075-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 187.0 to 191.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR