Ketorolac tromethamine ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

K0053-1G K0053-5G
TCK0053-1GEA 117.39 CAD
TCK0053-1G TCK0053-5G
Ketorolac tromethamine ≥98.0% (by HPLC, titration analysis)
Ketorolac tromethamine
Formula: C₁₅H₁₃NO₃·C₄H₁₁NO₃
MDL Number: MFCD00887595
CAS Number: 74103-07-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,05306

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR