Cyclobenzaprine hydrochloride ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Danger

C3157-1G C3157-5G
TCC3157-1GEA 58.46 CAD
TCC3157-1G TCC3157-5G
Cyclobenzaprine hydrochloride ≥98.0% (by HPLC, titration analysis)
Cyclobenzaprine hydrochloride
Formula: C₂₀H₂₁N·HCl
MDL Number: MFCD00079039
CAS Number: 6202-23-9
EINECS: 228-264-4
UN: 2811
ADR: 6.1,III
Merck Index: 14,02713

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 215.0 to 219.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR