Graphene, nanoplatelets aggregates, sub-micronparticles surface area 750 m²/g

Supplier: TCI America
Warning

G0502-25G G0502-5G
TCG0502-25GEA 289.3 CAD
TCG0502-25G TCG0502-5G
Graphene, nanoplatelets aggregates, sub-micronparticles surface area 750 m²/g
Graphene
MDL Number: MFCD00144065
CAS Number: 1034343-98-0

Order Now

Specification Test Results

Appearance Black powder to crystal
Specific surface 710 to 790 m2/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR