Graphene, nanoplatelets aggregates, sub-micronparticles surface area 500 m²/g

Supplier: TCI America
Warning

G0501-25G G0501-5G
TCG0501-25GEA 584.6 CAD
TCG0501-25G TCG0501-5G
Graphene, nanoplatelets aggregates, sub-micronparticles surface area 500 m²/g
Graphene
MDL Number: MFCD00144065
CAS Number: 1034343-98-0

Order Now

Specification Test Results

Appearance Black powder to crystal
Specific surface 470 to 530 m2/g


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR