Graphene, nanoplatelets, Length: 5 µm, Ø 2-10 nm

Supplier: TCI America
Warning

G0499-1G G0499-5G
TCG0499-1GEA 200.66 CAD
TCG0499-1G TCG0499-5G
Graphene, nanoplatelets, Length: 5 µm, Ø 2-10 nm
Graphene
MDL Number: MFCD00144065
CAS Number: 1034343-98-0

Order Now

Specification Test Results

Appearance Light gray to Brown to Black powder to crystal


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR