Graphene, nanoplatelets, Length: 15 µm, Ø 6-8 nm

Supplier: TCI America
Warning

G0442-25G G0442-5G
TCG0442-25GEA 508.76 CAD
TCG0442-25G TCG0442-5G
Graphene, nanoplatelets, Length: 15 µm, Ø 6-8 nm
Graphene
MDL Number: MFCD00144065
CAS Number: 1034343-98-0

Order Now

Specification Test Results

Appearance Black powder to crystal


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR