Graphene, nanoplatelets, Length: 5 µm, Ø 6-8 nm

Supplier: TCI America
Warning

G0441-25G G0441-5G
TCG0441-25GEA 508.76 CAD
TCG0441-25G TCG0441-5G
Graphene, nanoplatelets, Length: 5 µm, Ø 6-8 nm
Graphene
MDL Number: MFCD00144065
CAS Number: 1034343-98-0

Order Now

Specification Test Results

Appearance Black powder to lump
Thick 6 to 8 nm
Average size Equal to 5 micro_m


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR