5-Fluoro-2-hydroxybenzeneboronic acid (contains varying amounts of Anhydride)

Supplier: TCI America
Warning

F0977-1G F0977-5G
TCF0977-1GEA 107.44 CAD
TCF0977-1G TCF0977-5G
5-Fluoro-2-hydroxybenzeneboronic acid (contains varying amounts of Anhydride)
5-Fluoro-2-hydroxybenzeneboronic acid
Formula: C₆H₆BFO₃
MDL Number: MFCD03095357
CAS Number: 259209-20-6

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) 97.0 to 114.0 %

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR