2,3-Dihydrothieno[3,4-b]-1,4-dioxin-5-carbaldehyde ≥98.0% (by GC)

Supplier: TCI America

E1150-1G E1150-5G
TCE1150-1GEA 94.8 CAD
TCE1150-1G TCE1150-5G
2,3-Dihydrothieno[3,4-b]-1,4-dioxin-5-carbaldehyde ≥98.0% (by GC)
2,3-Dihydrothieno[3,4-b]-1,4-dioxin-5-carbaldehyde
Formula: C₇H₆O₃S
MDL Number: MFCD01651766
CAS Number: 204905-77-1

Order Now

Specification Test Results

Purity (GC) min. 98.0 %
Melting point 139.0 to 143.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR