2-Oxo-2H-chromen-7-yltrifluormethanesulfonate ≥98.0% (by GC)

Supplier: TCI America
Warning

O0472-1G O0472-5G
TCO0472-1GEA 350.76 CAD
TCO0472-1G TCO0472-5G
2-Oxo-2H-chromen-7-yltrifluormethanesulfonate ≥98.0% (by GC)
2-Oxo-2H-chromen-7-yltrifluormethanesulfonate
Formula: C₁₀H₅F₃O₅S
MDL Number: MFCD18382236
CAS Number: 108530-10-5

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 81.0 to 85.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR