2-Bromo-6-phenylpyridine ≥98.0% (by GC)

Supplier: TCI America
Warning

B5150-1G B5150-200MG
TCB5150-1GEA 271.76 CAD
TCB5150-1G TCB5150-200MG
2-Bromo-6-phenylpyridine ≥98.0% (by GC)
2-Bromo-6-phenylpyridine
Formula: C₁₁H₈BrN
MDL Number: MFCD00234691
CAS Number: 39774-26-0

Order Now

Specification Test Results

Appearance White to Yellow to Green powder to lump
Purity (GC) min. 98.0 %
Melting point 50.0 to 54.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR