Methyl-4-methoxy-2-methylbenzoate ≥98.0% (by GC)

Supplier: TCI America

M2889-1G M2889-5G
TCM2889-1GEA 61.62 CAD
TCM2889-1G TCM2889-5G
Methyl-4-methoxy-2-methylbenzoate ≥98.0% (by GC)
Methyl-4-methoxy-2-methylbenzoate
Formula: C₁₀H₁₂O₃
Boiling Pt: 98°C
MDL Number: MFCD06203918
CAS Number: 35598-05-1

Order Now

Specification Test Results

Appearance White to Light yellow clear liquid
Purity (GC) min. 98.0 %
Acid Value max. 3.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR