2-Ethyl-5-nitroaniline ≥98.0% (by GC)

Supplier: TCI America
Warning

E1085-1G E1085-5G
TCE1085-1GEA 173.8 CAD
TCE1085-1G TCE1085-5G
2-Ethyl-5-nitroaniline ≥98.0% (by GC)
2-Ethyl-5-nitroaniline
Formula: C₈H₁₀N₂O₂
MDL Number: MFCD00034056
CAS Number: 20191-74-6

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 96.0 %
Melting point 58.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR