1-(3-Chloropropyl)piperidine hydrochloride ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Warning

C2922-1G C2922-5G
TCC2922-1GEA 59.57 CAD
TCC2922-1G TCC2922-5G
1-(3-Chloropropyl)piperidine hydrochloride ≥98.0% (by titrimetric analysis)
1-(3-Chloropropyl)piperidine hydrochloride
Formula: C₈H₁₇Cl₂N
MDL Number: MFCD00012838
CAS Number: 5472-49-1
EINECS: 226-812-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.0 %
Purity (Argentmetric Titration) min. 98.0 %
Melting point 216.0 to 220.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR