Decahydroisoquinoline cis- and trans-mixture ≥98.0% (by GC)

Supplier: TCI America
Warning

D5067-25ML D5067-5ML
TCD5067-25MLEA 837.4 CAD
TCD5067-25ML TCD5067-5ML
Decahydroisoquinoline cis- and trans-mixture ≥98.0% (by GC)
Decahydroisoquinoline
Formula: C₉H₁₇N
Boiling Pt: 80°C
Flash Pt: 79°C
MDL Number: MFCD00012096
CAS Number: 6329-61-9
EINECS: 228-702-4

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 % (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR