2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 2-Hydrazinyl-4,5-dihydro-1H-imidazole hydrobromide, 2-Hydrazino-2-imidazoline Hydrobromide, 2-Hydrazino-4,5-dihydroimidazole Hydrobromide

H1523-1G H1523-5G
TCH1523-1GEA 121.66 CAD
TCH1523-1G TCH1523-5G
2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide ≥98.0% (by HPLC, titration analysis)
2-Hydrazino-4,5-dihydro-1H-imidazole hydrobromide
Formula: C₃H₉BrN₄
MW: 181.04 g/mol
MDL Number: MFCD00012694
CAS Number: 55959-84-7
EINECS: 259-921-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Potassium iodate Method) min. 98.0 %
Melting point 183.0 to 187.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR