(2-Thienylethynyl)trimethylsilane ≥98.0% (by GC)

Supplier: TCI America
Warning

T3189-1G T3189-5G
TCT3189-1GEA 164.32 CAD
TCT3189-1G TCT3189-5G
(2-Thienylethynyl)trimethylsilane ≥98.0% (by GC)
(2-Thienylethynyl)trimethylsilane
Formula: C₉H₁₂SSi
Boiling Pt: 90°C
Flash Pt: 25°C
MDL Number: MFCD03427125
CAS Number: 40231-03-6
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR