Ethyl-2-acetyl-3-ethoxyacrylate ≥98.0% (by GC)

Supplier: TCI America

Synonyms: Ethyl 2-(ethoxymethylene)acetoacetate, Ethyl 2-(ethoxymethylene)-3-oxobutanoate

E1148-25G E1148-5G
TCE1148-25GEA 202.24 CAD
TCE1148-25G TCE1148-5G
Ethyl-2-acetyl-3-ethoxyacrylate ≥98.0% (by GC)
Ethyl-2-acetyl-3-ethoxyacrylate
Formula: C₉H₁₄O₄
MW: 186.21 g/mol
Boiling Pt: 266°C
MDL Number: MFCD09959866
CAS Number: 3788-94-1
EINECS: 223-259-3

Order Now

Specification Test Results

Appearance Light yellow to Brown clear liquid
Purity (GC) min. 98.0 % (total of isomer)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR