Ethyl pyrimidine-4-carboxylate ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

E0881-25G E0881-5G
TCE0881-25GEA 662.81 CAD
TCE0881-25G TCE0881-5G
Ethyl pyrimidine-4-carboxylate ≥98.0% (by GC, titration analysis)
Ethyl pyrimidine-4-carboxylate
Formula: C₇H₈N₂O₂
Boiling Pt: 160°C
MDL Number: MFCD09832999
CAS Number: 62846-82-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to lump
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 37.0 to 41.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR