6-Fluorochromane-2-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 6-Fluorochroman-2-carboxylic acid, 6-Fluoro-3,4-dihydro-2H-1-benzopyran-2-carboxylic acid

F1086-25G F1086-5G
TCF1086-25GEA 549.84 CAD
TCF1086-25G TCF1086-5G
6-Fluorochromane-2-carboxylic acid ≥98.0% (by GC, titration analysis)
6-Fluorochromane-2-carboxylic acid
Formula: C₁₀H₉FO₃
MDL Number: MFCD07778357
CAS Number: 99199-60-7

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 124.0 to 128.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR