(7-Methoxy-1-naphthyl)acetic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

M2888-1G M2888-5G
TCM2888-1GEA 148.52 CAD
TCM2888-1G TCM2888-5G
(7-Methoxy-1-naphthyl)acetic acid ≥98.0% (by GC, titration analysis)
(7-Methoxy-1-naphthyl)acetic acid
Formula: C₁₃H₁₂O₃
MDL Number: MFCD12756806
CAS Number: 6836-22-2

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 155.0 to 159.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR