2-Amino-4-chloro-5-formylthiophene-3-carbonitrile ≥98.0% (by GC)

Supplier: TCI America
Danger

A2863-1G A2863-5G
TCA2863-1GEA 110.6 CAD
TCA2863-1G TCA2863-5G
2-Amino-4-chloro-5-formylthiophene-3-carbonitrile ≥98.0% (by GC)
2-Amino-4-chloro-5-formylthiophene-3-carbonitrile
Formula: C₆H₃ClN₂OS
Storage Temperature: Refrigerator
MDL Number: MFCD03788265
CAS Number: 104366-23-6
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Appearance Light yellow to Brown powder to crystal
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR