1,1'-Biphenyl-2,2'-diamine ≥99.0% (by GC, titration analysis)
Supplier: TCI America
|
Warning
|
D5255-1G
D5255-5G
76219-244EA
210.14
CAD
76219-244
76219-246
1,1'-Biphenyl-2,2'-diamine ≥99.0% (by GC, titration analysis)
1,1'-Biphenyl-2,2'-diamine
Formula:
C₁₂H₁₂N₂ Boiling Pt: 151°C |
MDL Number:
MFCD00034060 CAS Number: 1454-80-4 |
Specification Test Results
Appearance | White to Brown powder to crystal |
Purity (GC) | min. 99.0 % |
Purity (Nonaqueous Titration) | min. 99.0 % |
Melting point | 76.0 to 80.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...