Levoglucosenone ≥96.0% (by GC)

Supplier: TCI America

L0150-1G L0150-200MG
TCL0150-1GEA 837.4 CAD
TCL0150-1G TCL0150-200MG
Levoglucosenone ≥96.0% (by GC)
Levoglucosenone
Formula: C₆H₆O₃
Boiling Pt: 113°C
MDL Number: MFCD00191664
CAS Number: 37112-31-5
Merck Index: 14,05465

Order Now

Specification Test Results

Appearance White to Yellow to Orange clear liquid
Purity (GC) min. 96.0 %
Specific rotation [a]20/D -530 to -570 deg (C=1, CHCl3)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR