1-Bromo-4-methoxybutane ≥98.0% (by GC)

Supplier: TCI America
Warning

B4495-25G B4495-5G
TCB4495-25GEA 233.84 CAD
TCB4495-25G TCB4495-5G
1-Bromo-4-methoxybutane ≥98.0% (by GC)
1-Bromo-4-methoxybutane
Formula: C₅H₁₁BrO
Boiling Pt: 165°C
MDL Number: MFCD00037102
CAS Number: 4457-67-4
UN: 1993
ADR: 3,III

Order Now

Specification Test Results

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR