3-Chloro-4-((3-fluorobenzyl)oxy)aniline ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

C2874-1G C2874-5G
TCC2874-1GEA 58.46 CAD
TCC2874-1G TCC2874-5G
3-Chloro-4-((3-fluorobenzyl)oxy)aniline ≥98.0% (by GC, titration analysis)
3-Chloro-4-((3-fluorobenzyl)oxy)aniline
Formula: C₁₃H₁₁ClFNO
MDL Number: MFCD06809822
CAS Number: 202197-26-0
EINECS: 445-590-4
UN: 3077
ADR: 9,III

Order Now

Specification Test Results

Appearance White to Gray to Red powder to crystal
Purity (GC) min. 98.0 %
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 78.0 to 82.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR