Pemetrexed ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: (S)-2-(4-(2-(2-Amino-4-oxo-4,7-dihydro-1H-pyrrolo-[2,3-d]pyrimidin-5-yl)ethyl)benzamido)pentanedioic acid, (S)-2-(4-(2-(2-Amino-4-oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl)benzamido)pentanedioic acid, Alimta, [3H]-, N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid

P2288-1G P2288-250MG
TCP2288-1GEA 317.58 CAD
TCP2288-1G TCP2288-250MG
Pemetrexed ≥98.0% (by HPLC, titration analysis)
Pemetrexed
Formula: C₂₀H₂₁N₅O₆
Storage Temperature: Refrigerator
MDL Number: MFCD19105065
CAS Number: 137281-23-3
Merck Index: 14,07077

Order Now

Specification Test Results

Appearance White to Blue powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water 2.0 to 11.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR