(S)-N,N-Dimethyl-3-(naphthalen-1-yloxy)-1-phenylpropan-1-amine hydrochloride ≥98.0% (by HPLC)

Supplier: TCI America
Warning

D4761-1G D4761-5G
TCD4761-1GEA 88.48 CAD
TCD4761-1G TCD4761-5G
(S)-N,N-Dimethyl-3-(naphthalen-1-yloxy)-1-phenylpropan-1-amine hydrochloride ≥98.0% (by HPLC)
(S)-N,N-Dimethyl-3-(naphthalen-1-yloxy)-1-phenylpropan-1-amine hydrochloride
Formula: C₂₁H₂₄ClNO
Storage Temperature: Refrigerator
MDL Number: MFCD08272809
CAS Number: 129938-20-1
Merck Index: 14,02821

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 97.0 %
Melting point 179.0 to 183.0 deg-C
Specific rotation [a]20/D +123 to +128 deg (C=1, MeOH)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR