3,4,5-Trichlorophenylboronic acid (contains varying amounts of Anhydride)
Supplier: TCI America
|
Warning
|
T3383-1G
T3383-5G
TCT3383-1GEA
140.62
CAD
TCT3383-1G
TCT3383-5G
3,4,5-Trichlorophenylboronic acid (contains varying amounts of Anhydride)
3,4,5-Trichlorophenylboronic acid
Formula:
C₆H₄BCl₃O₂ |
MDL Number:
MFCD03095126 CAS Number: 862248-93-9 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (Neutralization titration) | 98.0 to 109.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...