4-Ethoxyphenylacetylene ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
1-Ethoxy-4-ethynylbenzene, 4-Ethoxyphenylacetylene
E1029-1G
E1029-5G
TCE1029-1GEA
52.14
CAD
TCE1029-1G
TCE1029-5G
4-Ethoxyphenylacetylene ≥98.0% (by GC)
4-Ethoxyphenylacetylene
Formula:
C₁₀H₁₀O MW: 146.19 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00173880 CAS Number: 79887-14-2 |
Specification Test Results
Appearance | Light orange to Yellow to Green clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...