4-Amino-1-methyl-3-n-propyl-1H-pyrazole-5-carboxamide ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 4-Amino-1-methyl-3-N-propyl-1H-pyrazole-5-carboxamide, 4-Amino-1-methyl-3-propylpyrazole-5-carboxamide

A2857-25G A2857-5G
TCA2857-25GEA 461.36 CAD
TCA2857-25G TCA2857-5G
4-Amino-1-methyl-3-n-propyl-1H-pyrazole-5-carboxamide ≥98.0% (by HPLC, titration analysis)
4-Amino-1-methyl-3-n-propyl-1H-pyrazole-5-carboxamide
Formula: C₈H₁₄N₄O
MW: 182.23 g/mol
MDL Number: MFCD02927682
CAS Number: 139756-02-8

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Nonaqueous Titration) min. 98.0 %
Melting point 98.0 to 102.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR