5-Bromobenzofuran ≥97.0% (by GC)
Supplier: TCI America
|
Warning
|
Synonyms:
5-Bromobenzo[b]furan
B3889-1G
B3889-5G
TCB3889-1GEA
79.02
CAD
TCB3889-1G
TCB3889-5G
5-Bromobenzofuran ≥97.0% (by GC)
5-Bromobenzofuran
Formula:
C₈H₅BrO Boiling Pt: 226°C Melting Pt: 8°C |
MDL Number:
MFCD03407317 CAS Number: 23145-07-5 |
Specification Test Results
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 97.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...