N4-(3-Bromophenyl)quinazoline-4,6-diamine ≥98.0% (by HPLC)
Supplier: TCI America
|
Warning
|
B5416-100MG
B5416-25MG
TCB5416-100MGEA
230.68
CAD
TCB5416-100MG
TCB5416-25MG
N4-(3-Bromophenyl)quinazoline-4,6-diamine ≥98.0% (by HPLC)
N4-(3-Bromophenyl)quinazoline-4,6-diamine
Formula:
C₁₄H₁₁BrN₄ Storage Temperature: Refrigerator |
MDL Number:
MFCD09908082 CAS Number: 169205-78-1 |
Specification Test Results
Appearance | Light yellow to Amber to Dark green powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Elemental analysis (Nitrogen) | 16.50 to 18.50 % |
Melting point | 202.0 to 206.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...