4-(3,4-Dichlorophenyl)-1-tetralone ≥98.0% (by GC)

Supplier: TCI America

Synonyms: 4-(3,4-Dichlorophenyl)-3,4-dihydronaphthalen-1(2H)-one

D4766-1G D4766-5G
TCD4766-1GEA 74.26 CAD
TCD4766-1G TCD4766-5G
4-(3,4-Dichlorophenyl)-1-tetralone ≥98.0% (by GC)
4-(3,4-Dichlorophenyl)-1-tetralone
Formula: C₁₆H₁₂Cl₂O
MW: 291.18 g/mol
MDL Number: MFCD02093088
CAS Number: 79560-19-3

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 104.0 to 108.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR