Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate ≥98.0% (by HPLC, titration analysis)

Supplier: TCI America

E1123-1G E1123-5G
TCE1123-1GEA 121.66 CAD
TCE1123-1G TCE1123-5G
Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate ≥98.0% (by HPLC, titration analysis)
Ethyl-4-oxo-3,4-dihydro-2-quinazolinecarboxylate
Formula: C₁₁H₁₀N₂O₃
MDL Number: MFCD00051583
CAS Number: 29113-33-5

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 %
Melting point 190.0 to 194.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR