1-Methyl-1H-pyrazole-3-carboxylic acid ≥97.0% (by GC, titration analysis)

Supplier: TCI America
Warning

Synonyms: 1-Methylpyrazole-3-carboxylic acid

M2642-1G M2642-200MG
TCM2642-1GEA 121.66 CAD
TCM2642-1G TCM2642-200MG
1-Methyl-1H-pyrazole-3-carboxylic acid ≥97.0% (by GC, titration analysis)
1-Methyl-1H-pyrazole-3-carboxylic acid
Formula: C₅H₆N₂O₂
MW: 126.12 g/mol
MDL Number: MFCD00464254
CAS Number: 25016-20-0

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 97.0 %
Purity (Neutralization titration) min. 97.0 %
Melting point 153.0 to 157.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR