4-Bromothiophene-2-methanol ≥98.0% (by GC)
Supplier: TCI America
Synonyms:
4-Bromo-2-thiophenemethanol, (4-Bromothiophen-2-yl)methanol, 4-Bromo-2-(hydroxymethyl)thiophene
B5060-1G
B5060-5G
TCB5060-1GEA
63.2
CAD
TCB5060-1G
TCB5060-5G
4-Bromothiophene-2-methanol ≥98.0% (by GC)
4-Bromothiophene-2-methanol
Formula:
C₅H₅BrOS |
MDL Number:
MFCD04115392 CAS Number: 79757-77-0 |
Specification Test Results
Appearance | White to Orange to Green powder to lump |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...