1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America
Warning

E1228-1G E1228-5G
76220-034EA 71.1 CAD
76220-034 76220-036
1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid ≥98.0% (by GC, titration analysis)
1-Ethyl-3-methyl-1H-pyrazole-5-carboxylic acid
Formula: C₇H₁₀N₂O₂
MDL Number: MFCD00156147
CAS Number: 50920-65-5

Order Now

Specification Test Results

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 142.0 to 146.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR