1-(4-Fluorophenyl)-1H-pyrrole-2,5-dione ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
F1054-1G
F1054-200MG
TCF1054-1GEA
279.66
CAD
TCF1054-1G
TCF1054-200MG
1-(4-Fluorophenyl)-1H-pyrrole-2,5-dione ≥98.0% (by GC)
1-(4-Fluorophenyl)-1H-pyrrole-2,5-dione
Formula:
C₁₀H₆FNO₂ |
MDL Number:
MFCD00033653 CAS Number: 6633-22-3 |
Specification Test Results
Appearance | White to Yellow to Green powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 154.0 to 158.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...