1-(1,3-Thiazol-2-yl)propan-1-one ≥98.0% (by GC)

Supplier: TCI America

P2247-25G P2247-5G
TCP2247-25GEA 398.16 CAD
TCP2247-25G TCP2247-5G
1-(1,3-Thiazol-2-yl)propan-1-one ≥98.0% (by GC)
1-(1,3-Thiazol-2-yl)propan-1-one
Formula: C₆H₇NOS
Boiling Pt: 107°C
MDL Number: MFCD01681404
CAS Number: 43039-98-1

Order Now

Specification Test Results

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR