4-Bromo-2-methylindan-1-one ≥98.0% (by GC)

Supplier: TCI America
Warning

B5096-1G B5096-5G
TCB5096-1GEA 83.74 CAD
TCB5096-1G TCB5096-5G
4-Bromo-2-methylindan-1-one ≥98.0% (by GC)
4-Bromo-2-methylindan-1-one
Formula: C₁₀H₉BrO
Boiling Pt: 134°C
MDL Number: MFCD09881709
CAS Number: 174702-59-1

Order Now

Specification Test Results

Appearance White or Colorless to Light yellow powder to lump to clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR