Dibenzo[b,d]furan-4-carbaldehyde ≥98.0% (by GC)

Supplier: TCI America
Warning

D4771-1G D4771-5G
TCD4771-1GEA 120.08 CAD
TCD4771-1G TCD4771-5G
Dibenzo[b,d]furan-4-carbaldehyde ≥98.0% (by GC)
Dibenzo[b,d]furan-4-carbaldehyde
Formula: C₁₃H₈O₂
MDL Number: MFCD03306027
CAS Number: 96706-46-6

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 98.0 %
Melting point 95.0 to 99.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR