3-(Bromoacetyl)-2H-chromen-2-one ≥95.0% (by GC)

Supplier: TCI America
Danger

B4679-1G B4679-5G
TCB4679-1GEA 45.82 CAD
TCB4679-1G TCB4679-5G
3-(Bromoacetyl)-2H-chromen-2-one ≥95.0% (by GC)
3-(Bromoacetyl)-2H-chromen-2-one
Formula: C₁₁H₇BrO₃
MDL Number: MFCD00488331
CAS Number: 29310-88-1

Order Now

Specification Test Results

Appearance White to Orange to Green powder to crystal
Purity (GC) min. 95.0 %
Melting point 163.0 to 167.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR