3-Ethoxycarbonylcoumarin ≥98.0% (by GC)
Supplier: TCI America
|
Warning
|
E0992-25G
E0992-5G
TCE0992-25GEA
350.44
CAD
TCE0992-25G
TCE0992-5G
3-Ethoxycarbonylcoumarin ≥98.0% (by GC)
3-Ethoxycarbonylcoumarin
Formula:
C₁₂H₁₀O₄ |
MDL Number:
MFCD00016964 CAS Number: 1846-76-0 |
Specification Test Results
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Melting point | 92.0 to 96.0 deg-C |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...