PYREX® Serological Pipettes, Corning
Supplier: Corning
7078B-50 7078-1CN 7078-1N 7078-2N 7078-5N 7078-10N 7077-1N 7077-2N 7077-5N 7077-10N 7077B-1 7077B-5 7077B-10 7077B-25 7077B-50 7079-1N 7079-2N 7079-5N 7079-10N 7078D-10 7078B-10 7078D-1 7078B-1 7078B-25 7078D-5 7078B-5
53224-610CS 606.69 CAD
53224-610 53284-100 53284-121 53284-143 53284-165 53284-187 53284-325 53284-347 53284-369 53284-380 53284-391 53284-427 53284-449 53284-460 53284-463 53284-562 53284-584 53284-609 53284-620 89091-106 89091-120 89091-220 89091-224 89091-268 89091-484 89091-488
PYREX® Serological Pipettes, Corning
Pipettes Serological Pipettes
These sterile disposable 1 ml PYREX® serological pipettes, calibrated (TD) with blow-out, offer long, slender, tapered tips to make pipetting go faster and easier.
  • All pipettes have negative graduations
  • Fire-polished tips for burr-free, even flow rates
Easy-open Steri-View™ wrappers (one side paper, one side plastic) have color-coded sizes printed directly on the paper for easy identification by size.
Certifications: Colors conform to I.S.O. standards.
Packaging: Non sterile pipettes available in bulk package; sterile pipettes available inMore Product Information