1-Benzyl-3-phenyl-2-thiourea ≥98.0% (by HPLC, total nitrogen)

Supplier: TCI America
Danger

Synonyms: 1-Benzyl-3-phenylthiourea

B4997-1G B4997-5G
TCB4997-1GEA 188.02 CAD
TCB4997-1G TCB4997-5G
1-Benzyl-3-phenyl-2-thiourea ≥98.0% (by HPLC, total nitrogen)
1-Benzyl-3-phenyl-2-thiourea
Formula: C₁₄H₁₄N₂S
MW: 242.35 g/mol
MDL Number: MFCD00022120
CAS Number: 726-25-0
UN: 2811
ADR: 6.1 III

Order Now

Specification Test Results

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (with Total Nitrogen) min. 98.0 %
Melting point 153.0 to 157.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR